Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzylaniline, 98%

$83.27 - $261.68

Chemical Identifiers

Chemical Name or Material 3-Benzylaniline
CAS 61424-26-8
Molecular Formula C13H13N
Formula Weight 183.25
MDL Number MFCD01310782
Beilstein 2803261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1987203

  SDS
alfa aesar™
L1987203
1g Each for $83.27

AAL1987206

  SDS
alfa aesar™
L1987206
5g Each for $261.68
Description & Specifications

Specifications

Formula Weight 183.25
Chemical Name or Material 3-Benzylaniline
Assay 98%
Molecular Formula C13H13N
CAS 61424-26-8
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD01310782
Beilstein 2803261
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 126°C (2mmHg)
Melting Point 48° to 52°C
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.