Please login to your online account to display your discounted pricing

3-(Benzylamino)propionitrile, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-(Benzylamino)propionitrile
Name Note 97%
CAS 706-03-6
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C10 H12 N2
Linear Formula C6H5CH2NHCH2CH2CN
Formula Weight 160.22
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC180601000

  SDS
thermo fisher scientific
180601000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 160.22
Assay ≥96.0%
Physical Form Liquid
Quantity 100g
Color Clear colorless
TSCA TSCA
EINECS Number 211-890-7
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Assay Percent Range 97%
MDL Number MFCD00001955
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Chemical Name or Material 3-(Benzylamino)propionitrile
CAS 706-03-6
Linear Formula C6H5CH2NHCH2CH2CN
Molecular Formula C10 H12 N2
Name Note 97%
Packaging Glass bottle
Refractive Index 1.5298 to 1.5318 (20°C, 589nm)