Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzylamino-1-propanol, 97%

$334.50 - $334.50

Chemical Identifiers

Chemical Name or Material 3-Benzylamino-1-propanol
CAS 4720-29-0
Molecular Formula C6H5CH2NH(CH2)3OH
Formula Weight 165.24
MDL Number MFCD00020622
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2666403

  SDS
alfa aesar™
H2666403
1g Each for $334.50
Description & Specifications

Specifications

Formula Weight 165.24
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Quantity 1g
CAS 4720-29-0
Assay 97%
Health Hazard 1 Warning
MDL Number MFCD00020622
Chemical Name or Material 3-Benzylamino-1-propanol
Molecular Formula C6H5CH2NH(CH2)3OH
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.