Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzoylthiophene-2-carboxylic acid, 96%

$116.77 - $626.81

Chemical Identifiers

Chemical Name or Material 3-Benzoylthiophene-2-carboxylic acid
CAS 30006-03-2
Molecular Formula C12H8O3S
Formula Weight 232.26
MDL Number MFCD00159542
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2804003

  SDS
alfa aesar™
H2804003
1g Each for $626.81

AAH28040MC

  SDS
alfa aesar™
H28040MC
100mg Each for $116.77

AAH28040MD

  SDS
alfa aesar™
H28040MD
250mg Each for $229.03
Description & Specifications

Specifications

Formula Weight 232.26
Molecular Formula C12H8O3S
Melting Point 150° to 153°C
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 96%
MDL Number MFCD00159542
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 30006-03-2
Quantity 1g
Chemical Name or Material 3-Benzoylthiophene-2-carboxylic acid
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.