missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzoylpyridine 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 105811000

 View more versions of this product

Catalog No. AC105811000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
183.21
White to light yellow
96.0
3-Benzoylpyridine
97%
5424-19-1
MFCD00006394
Glass bottle
Crystalline Solid
100g
36° to 40°C
100.0
C6H5CO(C5H4N)
97%
C12H9NO
Authentic
Phenyl-3-pyridyl ketone

Chemical Identifiers

5424-19-1
100.0
C12H9NO
183.21
Phenyl-3-pyridyl ketone
≥96.0%
96.0
97%
C6H5CO(C5H4N)
MFCD00006394
3-Benzoylpyridine, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 226-561-3

RTECSNumber : OB6401000

TSCA : TSCA