Please login to your online account to display your discounted pricing

3-Benzoylpyridine 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC105810050

 View more versions of this product

Catalog No. AC105810050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Assay ≥96.0%
Formula Weight 183.21
Quantity 5g
Color White to light yellow
EINECS Number 226-561-3
TSCA TSCA
Melting Point 36° to 40°C
RTECS Number OB6401000
CAS Min % 96.0
CAS Max % 100.0
CAS 5424-19-1
Synonym Phenyl-3-pyridyl ketone
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 3-Benzoylpyridine
Name Note 97%
Assay Percent Range 97%
Molecular Formula C12H9NO
Linear Formula C6H5CO(C5H4N)
MDL Number MFCD00006394
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

Specifications

Chemical Name or Material 3-Benzoylpyridine
Name Note 97%
CAS 5424-19-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12H9NO
Linear Formula C6H5CO(C5H4N)
Formula Weight 183.21
MDL Number MFCD00006394
Synonym Phenyl-3-pyridyl ketone
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

SDS & Safety Documents