missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzoylpropionic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC105795000

 View more versions of this product

Catalog No. AC105795000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Benzoylpropionic acid
97.5
Crystalline Powder
98%
C6H5COCH2CH2CO2H
MFCD00002792
02,26; 03,19
Glass bottle
114° to 119°C
2051-95-8
100.0
≥97.5%
C10H10O3
178.19
10, 696
Authentic
White to cream
500g

Chemical Identifiers

2051-95-8
100.0
C10H10O3
178.19
3-Benzoylpropionic acid
97.5
98%
C6H5COCH2CH2CO2H
MFCD00002792
≥97.5%