missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Benzoylpropionic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 105790250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
3-Benzoylpropionic acid | |
97.5 | |
Authentic | |
Glass bottle | |
C6H5COCH2CH2CO2H | |
25g | |
02,26; 03,19 | |
KMQLIDDEQAJAGJ-UHFFFAOYSA-N | |
4-oxo-4-phenylbutanoic acid | |
72871 | |
178.19 |
2051-95-8 | |
100.0 | |
97.5% min. (HPLC) | |
C10H10O3 | |
MFCD00002792 | |
10, 696 | |
3-benzoylpropionic acid, 4-oxo-4-phenylbutyric acid, 3-benzoylpropanoic acid, beta-benzoylpropionic acid, benzoylpropionic acid, propanoic acid, 3-benzoyl, benzenebutanoic acid, .gamma.-oxo, 3-benzoylpropionicacid, propionic acid, 3-benzoyl, 4-phenyl-4-oxobutyric acid | |
OC(=O)CCC(=O)C1=CC=CC=C1 | |
178.19 | |
CHEBI:64437 | |
98% |
Safety and Handling
EINECSNumber : 218-135-
RUO â Research Use Only