missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Benzoylbenzoic acid, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals B2551914

 View more versions of this product

Catalog No. AAB2551914


Description

Description

3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Benzoylbenzoic acid has been used in light-induced reactions on benzophenone-terminated boron-doped diamond (BDD) surfaces and preparation of benzophenone flavonol derivative.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Specifications

3-Benzoylbenzoic acid
162°C to 166°C
C14H10O3
25 g
benzoic acid, 3-benzoyl, m-benzoylbenzoic acid, 3-benzoyl benzoic acid, 3-benzoyl-benzoic acid, 3-phenylcarbonyl benzoic acid, 3-benzoylbenzoicacid, 3-benzoyl-benzoesaure, 3-benzoylbenzoic-acid, acmc-20an9k, benzoic acid,3-benzoyl
AXJXRLHTQQONQR-UHFFFAOYSA-N
3-benzoylbenzoic acid
101386
98%
579-18-0
Odorless
MFCD00002518
2212366
Insoluble in water.
OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
226.23
226.23

Chemical Identifiers

579-18-0
226.23
AXJXRLHTQQONQR-UHFFFAOYSA-N
3-benzoylbenzoic acid
C14H10O3
MFCD00002518
101386
OC(=O)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only