missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzoylacrylic acid, predominantly trans, 97%

Manufacturer:  Alfa Aesar™ A1114714

 View more versions of this product

Catalog No. AAA1114714



Description

Description

Applications
2-Butenoic acid, 4-oxo-4-phenyl- could react with morpholine, and obtain the 2-morpholin-4-yl-4-oxo-4-phenyl-butyric acid. This reaction needs the solvent of toluene, ethanol.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

3-Benzoylacrylic acid
93°C to 97°C
predominantly trans
C10H8O3
UN2811
3-benzoylacrylic acid, benzoylacrylic acid, 4-oxo-4-phenyl-2-butenoic acid, e-4-oxo-4-phenylbut-2-enoic acid, acrylic acid, 3-benzoyl, 2-butenoic acid, 4-oxo-4-phenyl, 4-oxo-4-phenylbut-2-enoic acid, crotonic acid, 4-oxo-4-phenyl, .beta.-benzoylacrylic acid, trans-3-benzoylacrylic acid
C1=CC=C(C=C1)C(=O)C=CC(=O)O
176.171
176.17
Odorless
25g
583-06-2
MFCD00014015
1817876
PLPDHGOODMBBGN-VOTSOKGWSA-N
(E)-4-oxo-4-phenylbut-2-enoic acid
676116
97%

Chemical Identifiers

583-06-2
176.171
PLPDHGOODMBBGN-VOTSOKGWSA-N
676116
C1=CC=C(C=C1)C(=O)C=CC(=O)O
C10H8O3
MFCD00014015
3-benzoylacrylic acid, benzoylacrylic acid, 4-oxo-4-phenyl-2-butenoic acid, e-4-oxo-4-phenylbut-2-enoic acid, acrylic acid, 3-benzoyl, 2-butenoic acid, 4-oxo-4-phenyl, 4-oxo-4-phenylbut-2-enoic acid, crotonic acid, 4-oxo-4-phenyl, .beta.-benzoylacrylic acid, trans-3-benzoylacrylic acid
(E)-4-oxo-4-phenylbut-2-enoic acid
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : EM9259050

TSCA : TSCA

SDS Documents