Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Benzoyl-1,1,1-trifluoroacetone, 98+%

$187.80 - $845.07

Chemical Identifiers

Chemical Name or Material 3-Benzoyl-1,1,1-trifluoroacetone
CAS 326-06-7
Molecular Formula C10H7F3O2
Formula Weight 216.16
MDL Number MFCD00000425
Beilstein 1875083
Synonym 4,4,4-Trifluoro-1-phenyl-1,3-butanedione
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1711022

 
alfa aesar™
A1711022
100g Each for $187.80

AAA1711036

 
alfa aesar™
A1711036
500g Each for $845.07
Description & Specifications

Specifications

Formula Weight 216.16
Chemical Name or Material 3-Benzoyl-1,1,1-trifluoroacetone
TSCA TSCA
Assay ≥98%
Molecular Formula C10H7F3O2
Melting Point 37° to 40°C
Boiling Point 224°C
MDL Number MFCD00000425
Quantity 100g
CAS 326-06-7
Flash Point 98°C (208°F)
Beilstein 1875083
Synonym 4,4,4-Trifluoro-1-phenyl-1,3-butanedione

Applications
3-Benzoyl-1,1,1-trifluoroacetone, is used as a primary and secondary intermediate. It is used as an Intermediate in organic synthesis.

Solubility
Sparingly Soluble in water (0.24 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.