Learn More
3-Azido-1-propylamine, Thermo Scientific Chemicals
Azido-containing reagent with carbonyl and carboxyl reactivity
$228.48 - $1049.84
Chemical Identifiers
CAS | 88192-19-2 |
---|---|
Molecular Formula | C3H8N4 |
Molecular Weight (g/mol) | 100.125 |
MDL Number | MFCD11046568 |
InChI Key | OYBOVXXFJYJYPC-UHFFFAOYSA-N |
Synonym | 3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine |
PubChem CID | 150110 |
IUPAC Name | 3-azidopropan-1-amine |
SMILES | C(CN)CN=[N+]=[N-] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ65668MC
|
Thermo Scientific Chemicals
J65668MC |
100 mg |
Each for $228.48
|
|
|||||
AAJ6566803
|
Thermo Scientific Chemicals
J6566803 |
1 g |
Each for $1,049.84
|
|
|||||
Description
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsAzido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.
Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
88192-19-2 | |
100.125 | |
OYBOVXXFJYJYPC-UHFFFAOYSA-N | |
150110 | |
C(CN)CN=[N+]=[N-] |
C3H8N4 | |
MFCD11046568 | |
3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine | |
3-azidopropan-1-amine |
Specifications
88192-19-2 | |
60°C (140°F) | |
N3CH2CH2CH2NH2 | |
100 mg | |
3-azidopropylamine, 3-azido-1-propanamine, 1-propanamine, 3-azido, 3-azido-propylamine, 1-azido-3-aminopropane, 1-amino-3-azidopropane, 3-aminopropyl azide, 3-azido-1-propylamine, 3-azidopropyl-1-amine | |
OYBOVXXFJYJYPC-UHFFFAOYSA-N | |
3-azidopropan-1-amine | |
150110 | |
Liquid |
Yellow | |
C3H8N4 | |
MFCD11046568 | |
UN1993 | |
Soluble in DMSO,DMF,DCM,chloroform,THF,and water;Miscible with dimethylsulfoxide,dimethyl formamide,dichloromethane,tetrahydrofuran,chloroform and water. | |
C(CN)CN=[N+]=[N-] | |
100.125 | |
100.12 | |
3-Azido-1-propylamine |
Safety and Handling
GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only