missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Azetidinecarboxylic acid, 97%, ACROS Organics™

Chemical Identifiers

CAS 36476-78-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.105
MDL Number MFCD00191763
InChI Key GFZWHAAOIVMHOI-UHFFFAOYSA-N
Synonym 3-azetanecarboxylic acid
PubChem CID 93192
IUPAC Name azetidine-3-carboxylic acid
SMILES C1C(CN1)C(=O)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC369620500 SDS View Documents Thermo Fisher Scientific
AC369620500
50mg Glass Bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

36476-78-5
101.105
GFZWHAAOIVMHOI-UHFFFAOYSA-N
93192
C1C(CN1)C(=O)O
C4H7NO2
MFCD00191763
3-azetanecarboxylic acid
azetidine-3-carboxylic acid

Specifications

Authentic
White
36476-78-5
100.0
C4H7NO2
3-azetanecarboxylic acid
C1C(CN1)C(=O)O
101.105
101.11
≥96.0%
Glass Bottle
50mg
96.0
97%
MFCD00191763
GFZWHAAOIVMHOI-UHFFFAOYSA-N
azetidine-3-carboxylic acid
93192
Crystalline Powder
3-Azetidinecarboxylic acid