Please login to your online account to display your discounted pricing

3-Azetidinecarboxylic acid, 97%, ACROS Organics™


Quantity 50mg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC369620500

 View more versions of this product

Catalog No. AC369620500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 101.11
Quantity 50mg
Color White to off-white
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 3-Azetidinecarboxylic acid
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C4H7NO2
Packaging Glass bottle
MDL Number MFCD00191763
CAS 36476-78-5

Specifications

Chemical Name or Material 3-Azetidinecarboxylic acid
CAS 36476-78-5
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C4H7NO2
Formula Weight 101.11
MDL Number MFCD00191763