Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Azabicyclo[3.3.0]octane hydrochloride, 98%

Chemical Identifiers

Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride
CAS 112626-50-3
Formula Weight 147.65
MDL Number MFCD06411019
Beilstein 3672489
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6646214

 
alfa aesar™
H6646214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6646222

 
alfa aesar™
H6646222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.65
MDL Number MFCD06411019
Physical Form Solid
Assay 98%
CAS 112626-50-3
Chemical Name or Material 3-Azabicyclo[3.3.0]octane hydrochloride
Beilstein 3672489
Quantity 25g