missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-AQC, Tocris

5-HT3 antagonist

Manufacturer:  R & D Systems 066650

 View more versions of this product

Catalog No. 066650


 

Specifications Description & Specifications

Specifications

3-AQC
>98%
395.42
50mg
201216-42-4
C16H17N5·C4H4O4
3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate

Chemical Identifiers

>98%
395.42
3-AQC
C16H17N5·C4H4O4
3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate

Safety and Handling

Recommended Storage : Desiccate at -20°C