Please login to your online account to display your discounted pricing

3-AQC, Tocris


5-HT3 antagonist

Quantity 10mg

Manufacturer: r & d systems  066610

 View more versions of this product

Catalog No. 066610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 395.42
Molecular Formula C16H17N5·C4H4O4
Chemical Name or Material 3-AQC
Assay Percent Range >98%
Synonym 3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate
CAS 201216-42-4
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material 3-AQC
Assay Percent Range >98%
Molecular Formula C16H17N5·C4H4O4
Formula Weight 395.42
Synonym 3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate
Accessories

Desiccate at -20°C