Please login to your online account to display your discounted pricing

3-AQC, Tocris

5-HT3 antagonist

$181.19 - $762.58

Chemical Identifiers

Chemical Name or Material 3-AQC
Assay Percent Range >98%
Molecular Formula C16H17N5·C4H4O4
Formula Weight 395.42
Synonym 3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

066610

 
r & d systems
066610
10mg Each for $181.19

066650

 
r & d systems
066650
50mg Each for $762.58
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 395.42
Chemical Name or Material 3-AQC
Assay Percent Range >98%
Molecular Formula C16H17N5·C4H4O4
Synonym 3-(4-Allylpiperazin-1-yl)-2-quinoxalinecarbonitrile maleate
CAS 201216-42-4
Recommended Storage Desiccate at -20°C
Safety and Handling

Desiccate at -20°C