Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Anilinoacrolein anil, 98%

$59.51 - $59.51

Chemical Identifiers

Chemical Name or Material 3-Anilinoacrolein anil
CAS 4485-89-6
Molecular Formula C6H5NHCH=CHCH=NC6H5
Formula Weight 222.29
MDL Number MFCD00051805
Beilstein 609808
Synonym 3-Phenylamino-2-propenal N-phenylimine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0694703

  SDS
alfa aesar™
L0694703
1g Each for $59.51
Description & Specifications

Specifications

Formula Weight 222.29
Assay 98%
CAS 4485-89-6
Melting Point 111° to 113°C
Beilstein 609808
Chemical Name or Material 3-Anilinoacrolein anil
MDL Number MFCD00051805
Quantity 1g
Molecular Formula C6H5NHCH=CHCH=NC6H5
Synonym 3-Phenylamino-2-propenal N-phenylimine

Applications
3-Anilinoacrolein anil appears or found to be an equivalent to malondialdehyde in synthesis.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.