Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Anilino-2-cyclohexen-1-one, 98%

$86.66 - $335.57

Chemical Identifiers

Chemical Name or Material 3-Anilino-2-cyclohexen-1-one
CAS 24706-50-1
Molecular Formula C12H13NO
Formula Weight 187.24
MDL Number MFCD00082654
Beilstein 2103381
Synonym 3-(Phenylamino)cyclohex-2-ene-1-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1375006

 
alfa aesar™
A1375006
5g Each for $86.66

AAA1375014

 
alfa aesar™
A1375014
25g Each for $335.57
Description & Specifications

Specifications

Formula Weight 187.24
Chemical Name or Material 3-Anilino-2-cyclohexen-1-one
Assay 98%
Molecular Formula C12H13NO
MDL Number MFCD00082654
CAS 24706-50-1
Beilstein 2103381
Synonym 3-(Phenylamino)cyclohex-2-ene-1-one
Melting Point 176° to 178°C
Quantity 5g
RTECS Number GW7600000