Learn More
3-Aminothiophenol, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2547306
Description
3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminothiophenol was used as analyte during the fabrication of top-to-bottom joined system consisting of a silver nanowire and nanospheres. It was used in formation of polyaniline-analog monolayer on gold surface by anchoring to the gold substrate through the thiol moiety.
Solubility
Soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
3-Aminothiophenol | |
1.179 | |
>100°C (212°F) | |
1.659 | |
5 g | |
Air sensitive | |
Soluble in water. | |
NC1=CC=CC([S-])=C1 | |
124.18 | |
125.19 |
22948-02-3 | |
123°C to 125°C (8 mmHg) | |
C6H6NS | |
MFCD00007791 | |
UN3267 | |
3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c | |
KFFUEVDMVNIOHA-UHFFFAOYSA-M | |
3-aminobenzenethiol | |
31577 | |
97% |
Chemical Identifiers
22948-02-3 | |
124.18 | |
KFFUEVDMVNIOHA-UHFFFAOYSA-M | |
31577 |
C6H6NS | |
MFCD00007791 | |
3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c | |
NC1=CC=CC([S-])=C1 |
Safety and Handling
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P235-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H227-H302-H312-H314-H332-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
EINECSNumber : 245-344-4
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only