Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminothiophene-2-carboxamide, 97%

$63.14 - $185.34

Chemical Identifiers

Chemical Name or Material 3-Aminothiophene-2-carboxamide
CAS 147123-47-5
Molecular Formula C5H6N2OS
Formula Weight 142.18
MDL Number MFCD00052593
Beilstein 7165908
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1237103

  SDS
alfa aesar™
L1237103
1g Each for $185.34

AAL12371MD

  SDS
alfa aesar™
L12371MD
250mg Each for $63.14
Description & Specifications

Specifications

Formula Weight 142.18
Quantity 1g
Assay 97%
Molecular Formula C5H6N2OS
Melting Point 120° to 122°C
MDL Number MFCD00052593
CAS 147123-47-5
Beilstein 7165908
Chemical Name or Material 3-Aminothiophene-2-carboxamide