Please login to your online account to display your discounted pricing

3-Aminothieno[2,3-b]pyridine-2-carboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Aminothieno[2, 3-b]pyridine-2-carboxylic acid
CAS 58327-75-6
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H6N2O2S
Formula Weight 194.21
MDL Number MFCD03783349
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO00759EA

  SDS
fisher scientific company
MO00759EA
Amber Glass Bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 194.21
Quantity 10g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 3-Aminothieno[2, 3-b]pyridine-2-carboxylic acid
CAS 58327-75-6
Molecular Formula C8H6N2O2S
MDL Number MFCD03783349
Packaging Amber Glass Bottle
Assay Percent Range 97%
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.