Learn More
3-Aminoquinuclidine dihydrochloride, 98%, ACROS Organics™
$81.48 - $81.48
Chemical Identifiers
CAS | 6530-09-2 |
---|---|
Molecular Formula | C7H14N2·2ClH |
Molecular Weight (g/mol) | 199.12 |
MDL Number | MFCD00137395 |
InChI Key | STZHBULOYDCZET-UHFFFAOYNA-N |
Synonym | 1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, 3-aminoquinuclidine dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, quinuclidine-3-ylamine, chloride, chloride |
PubChem CID | 197853 |
IUPAC Name | 1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride |
SMILES | C1CN2CCC1C(C2)N.Cl.Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC146150050
|
Thermo Fisher Scientific
146150050 |
5g | Glass bottle |
Each for $81.48
|
|
AC146150250
|
Thermo Fisher Scientific
146150250 |
25g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
6530-09-2 | |
199.12 | |
STZHBULOYDCZET-UHFFFAOYNA-N | |
197853 | |
C1CN2CCC1C(C2)N.Cl.Cl |
C7H14N2·2ClH | |
MFCD00137395 | |
1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, 3-aminoquinuclidine dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, quinuclidine-3-ylamine, chloride, chloride | |
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride |
Specifications
Authentic | |
White | |
5g | |
97.5 | |
98% | |
MFCD00137395 | |
Solubility in water: soluble | |
C1CN2CCC1C(C2)N.Cl.Cl | |
199.12 | |
199.12 | |
98% |
Glass bottle | |
321°C to 323°C | |
6530-09-2 | |
100.0 | |
C7H14N2·2ClH | |
1-aza-bicyclo 2.2.2 oct-3-ylamine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine dihydrochloride, 1-azabicyclo 2.2.2 octan-3-amine, dihydrochloride, 3-amino-1-azabicyclo 2.2.2 octane dihydrochloride, 3-aminoquinuclidine dihydrochloride, 3aminoquinuclidine dihydrochloride, quinuclidin-3-amine dihydrochloride, quinuclidine, 3-amino-, dihydrochloride, quinuclidine-3-ylamine, chloride, chloride | |
STZHBULOYDCZET-UHFFFAOYNA-N | |
1-azabicyclo[2.2.2]octan-3-amine;dihydrochloride | |
197853 | |
Powder | |
3-Aminoquinuclidine dihydrochloride, 98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 229-424-6
RTECSNumber : VD4590000
TSCA : TSCA