Please login to your online account to display your discounted pricing

3-Aminoquinoline, 99+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC213780010

 View more versions of this product

Catalog No. AC213780010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder or Crystals
Assay ≥99.0%
Formula Weight 144.18
Quantity 1g
Color White to beige
EINECS Number 209-455-1
TSCA TSCA
Melting Point 92° to 96°C
RTECS Number VA9622000
Beilstein 22, I, 638
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material 3-Aminoquinoline
Name Note 99+%
Assay Percent Range 99+%
Molecular Formula C9H8N2
MDL Number MFCD00006772
CAS 580-17-6
Infrared Spectrum Authentic
Synonym 3-Quinolinamine
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause damage to organs.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Aminoquinoline
Name Note 99+%
CAS 580-17-6
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00006772
Synonym 3-Quinolinamine
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause damage to organs.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.
Harmful if inhaled.
Suspected of causing genetic defects.

SDS & Safety Documents