Learn More
3-Aminoquinoline, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 213780050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminoquinoline | |
580-17-6 | |
100.0 | |
White to Beige | |
98.5% min. (GC) | |
C9H8N2 | |
5 g | |
3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678 | |
SVNCRRZKBNSMIV-UHFFFAOYSA-N | |
quinolin-3-amine | |
11375 | |
99% |
99+% | |
99.0 | |
91.0°C to 94.0°C | |
Authentic | |
Glass bottle | |
MFCD00006772 | |
22, I, 638 | |
Solubility in water: slightly soluble in hot water. Other solubilities: soluble in methanol | |
C1=CC=C2C(=C1)C=C(C=N2)N | |
144.18 | |
144.18 | |
Crystalline Powder or Crystals |
Chemical Identifiers
580-17-6 | |
144.18 | |
SVNCRRZKBNSMIV-UHFFFAOYSA-N | |
11375 | |
C1=CC=C2C(=C1)C=C(C=N2)N |
C9H8N2 | |
MFCD00006772 | |
3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678 | |
quinolin-3-amine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi
GHS Signal Word: Warning
EINECSNumber : 209-455-1
RTECSNumber : VA9622000
TSCA : TSCA
RUO – Research Use Only