Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminoquinoline, 98%

$90.96 - $354.04

Chemical Identifiers

Chemical Name or Material 3-Aminoquinoline
CAS 580-17-6
Molecular Formula C9H8N2
Formula Weight 144.18
MDL Number MFCD00006772
Beilstein 113317
Synonym 3-Quinolinamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1659206

 
alfa aesar™
A1659206
5g Each for $90.96

AAA1659214

 
alfa aesar™
A1659214
25g Each for $354.04
Description & Specifications

Specifications

Formula Weight 144.18
Chemical Name or Material 3-Aminoquinoline
TSCA TSCA
Melting Point 92° to 96°C
Quantity 5g
Assay 98%
Molecular Formula C9H8N2
MDL Number MFCD00006772
CAS 580-17-6
Beilstein 113317
Synonym 3-Quinolinamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number VA9622000
Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P305+P351+P338-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.