Learn More
3-Aminoquinoline, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1659214
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminoquinoline | |
92°C to 96°C | |
MFCD00006772 | |
113317 | |
3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678 | |
C1=CC=C2C(=C1)C=C(C=N2)N | |
144.177 | |
144.18 |
580-17-6 | |
C9H8N2 | |
25 g | |
Air Sensitive | |
SVNCRRZKBNSMIV-UHFFFAOYSA-N | |
quinolin-3-amine | |
11375 | |
98% |
Chemical Identifiers
580-17-6 | |
144.177 | |
SVNCRRZKBNSMIV-UHFFFAOYSA-N | |
11375 | |
C1=CC=C2C(=C1)C=C(C=N2)N |
C9H8N2 | |
MFCD00006772 | |
3-aminoquinoline, 3-quinolinamine, 3-quinolylamine, 3-quinolineamine, quinoline, 3-amino, quinolin-3-ylamine, 3-amino quinoline, unii-98ovl5k3r4, quinolin-3-yl-amine, ccris 1678 | |
quinolin-3-amine |
Safety and Handling
GHS H Statement
H301-H311-H332-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 209-455-1
RTECSNumber : VA9622000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only