missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopyrrolidine dihydrochloride, 98%, ACROS Organics™

$91.95 - $91.95

Chemical Identifiers

CAS 103831-11-4
Molecular Formula C4H12Cl2N2
Molecular Weight (g/mol) 159.054
InChI Key NJPNCMOUEXEGBL-UHFFFAOYSA-N
Synonym 3-amino-pyrrolidine dihydrochloride, 3-aminopyrrolidine 2hcl, 3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, pyrrolidin-3-amine-hydrogen chloride 1/2, pyrrolidine-3-ylamine, chloride, chloride
PubChem CID 16212596
IUPAC Name pyrrolidin-3-amine;dihydrochloride
SMILES C1CNCC1N.Cl.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC345310050
SDS View Documents
Thermo Fisher Scientific
345310050
5g Glass bottle Each for $91.95
 
Specifications

Chemical Identifiers

103831-11-4
159.054
3-amino-pyrrolidine dihydrochloride, 3-aminopyrrolidine 2hcl, 3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, pyrrolidin-3-amine-hydrogen chloride 1/2, pyrrolidine-3-ylamine, chloride, chloride
pyrrolidin-3-amine;dihydrochloride
C4H12Cl2N2
NJPNCMOUEXEGBL-UHFFFAOYSA-N
16212596
C1CNCC1N.Cl.Cl

Specifications

Authentic
Cream to White
103831-11-4
100.0
C4H12Cl2N2
NJPNCMOUEXEGBL-UHFFFAOYSA-N
pyrrolidin-3-amine;dihydrochloride
16212596
Crystalline Powder
3-Pyrrolidinamine
Glass bottle
5g
97.5
98%
3-amino-pyrrolidine dihydrochloride, 3-aminopyrrolidine 2hcl, 3-aminopyrrolidine dihydrochloride, 3-pyrrolidinamine dihydrochloride, 3-pyrrolidinamine, dihydrochloride, ?-3-aminopyrrolidine dihydrochloride, pyrrolidin-3-amine dihydrochloride, pyrrolidin-3-amine-hydrogen chloride 1/2, pyrrolidine-3-ylamine, chloride, chloride
C1CNCC1N.Cl.Cl
159.054
86.14
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.