missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Aminopyridine, 99%, ACROS Organics™
$39.01 - $114.45
Chemical Identifiers
CAS | 462-08-8 |
---|---|
Molecular Formula | C5H6N2 |
Molecular Weight (g/mol) | 94.12 |
MDL Number | MFCD00006400 |
InChI Key | CUYKNJBYIJFRCU-UHFFFAOYSA-N |
Synonym | 3-amino pyridine, 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, amino-3 pyridine, amino-3-pyridine, beta-aminopyridine, m-aminopyridine, pyridin-3-ylamine, pyridine, 3-amino |
PubChem CID | 10009 |
IUPAC Name | pyridin-3-amine |
SMILES | C1=CC(=CN=C1)N |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC104560250
|
Thermo Fisher Scientific
104560250 |
25g | Glass bottle |
Each for $39.01
|
|
AC104561000
|
Thermo Fisher Scientific
104561000 |
100g | Glass bottle |
Each for $114.45
|
|
AC104565000
|
Thermo Fisher Scientific
104565000 |
500g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
462-08-8 | |
94.12 | |
CUYKNJBYIJFRCU-UHFFFAOYSA-N | |
10009 | |
C1=CC(=CN=C1)N |
C5H6N2 | |
MFCD00006400 | |
3-amino pyridine, 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, amino-3 pyridine, amino-3-pyridine, beta-aminopyridine, m-aminopyridine, pyridin-3-ylamine, pyridine, 3-amino | |
pyridin-3-amine |
Specifications
124°C | |
Glass bottle | |
248.0°C | |
60.0°C to 63.0°C | |
462-08-8 | |
100.0 | |
C5H6N2 | |
MFCD00006400 | |
15, 469 | |
Solubility in water: soluble. Other solubilities: soluble in alcohol, benzene and ether, insoluble in petroleum ether | |
C1=CC(=CN=C1)N | |
94.12 | |
94.12 | |
99% |
Authentic | |
1% max. (K.F.) | |
Colorless to Yellow | |
25g | |
98.5 | |
98.5% min. (GC) | |
(C5H4N)NH2 | |
22, 431 | |
3-amino pyridine, 3-aminopyridine, 3-pyridinamine, 3-pyridylamine, amino-3 pyridine, amino-3-pyridine, beta-aminopyridine, m-aminopyridine, pyridin-3-ylamine, pyridine, 3-amino | |
CUYKNJBYIJFRCU-UHFFFAOYSA-N | |
pyridin-3-amine | |
10009 | |
Liquid | |
3-Aminopyridine, 99% |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
GHS Signal Word: Danger
EINECSNumber : 207-322-2
RTECSNumber : US1650000
TSCA : TSCA