Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopyridine, 99%

Chemical Identifiers

Chemical Name or Material 3-Aminopyridine
CAS 462-08-8
Molecular Formula C5H6N2
Formula Weight 94.12
MDL Number MFCD00006400
UN Number UN2671
Beilstein 105692
Merck Index 14,473
Synonym 3-Pyridinamine; 3-Pyridylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA132160B

 
alfa aesar™
A132160B
1000g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 94.12
Chemical Name or Material 3-Aminopyridine
TSCA TSCA
Melting Point 58° to 63°C
Boiling Point 250° to 252°C
pH 10
Quantity 1000g
Flash Point 88°C (190°F)
Assay 99%
Odor Unpleasant
Molecular Formula C5H6N2
MDL Number MFCD00006400
CAS 462-08-8
UN Number UN2671
Beilstein 105692
Merck Index 14,473
Synonym 3-Pyridinamine; 3-Pyridylamine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280h-P270-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
Do not eat, drink or smoke when using this product.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: AMINOPYRIDINES
RTECS Number US1650000

Applications
3-Aminopyrodine is also used as an intermediate for agrochemicals, pharmaceuticals and colorants. These are involved in photosensitizer chemistry, luminescent materials, metal complex chemistry and liquid crystal. Aminopyridine is used as a monomer for polymerization. It is also used in the Production of cationic dyes and Apa amoxicillin sodium raw materials.

Solubility
It is soluble in water as well as soluble in alcohol, benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under ordinary conditions.

Safety and Handling

Danger

GHS P Statement
P280h-P270-P305+P351+P338-P309-P310
Wear protective gloves/clothing.
Do not eat, drink or smoke when using this product.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H311-H331-H315-H319-H335
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.