Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopyridine-4-carboxaldehyde, 95%

$100.51 - $344.80

Chemical Identifiers

Chemical Name or Material 3-Aminopyridine-4-carboxaldehyde
CAS 55279-29-3
Molecular Formula C6H6N2O
Formula Weight 122.13
MDL Number MFCD06410684
Synonym 3-Aminoisonicotinaldehyde
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5002403

 
alfa aesar™
H5002403
1g Each for $344.80

AAH50024MD

 
alfa aesar™
H50024MD
250mg Each for $100.51
Description & Specifications

Specifications

Formula Weight 122.13
Chemical Name or Material 3-Aminopyridine-4-carboxaldehyde
Melting Point ≈92°C
Quantity 1g
Assay 95%
Molecular Formula C6H6N2O
MDL Number MFCD06410684
CAS 55279-29-3
Synonym 3-Aminoisonicotinaldehyde
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Aminopyridine-4-carboxaldehyde is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.