missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopyridine-2-carboxylic acid, 98%

Manufacturer:  Alfa Aesar™ H6486114

 View more versions of this product

Catalog No. AAH6486114



Description

Description

Applications
Employed as an important intermediate for syntheses material and pharmaceutical.

Solubility
Soluble in acetone, ethanol and methanol.

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

3-Aminopyridine-2-carboxylic acid
218°C to 222°C
1462-86-8
MFCD00090153
2-pyridinecarboxylic acid, 3-amino, 2-pyridinecarboxylicacid, 3-amino, 3-amino-2-carboxypyridine, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-pyridine-2-carboxylic acid, 3-aminopicolinic acid, 3-aminopyridine-2-carboxylic acid, tech., pubchem2015
BOOMHTFCWOJWFO-UHFFFAOYSA-N
3-aminopyridine-2-carboxylic acid
73836
Solid
Brown
25g
C6H6N2O2
3600
Soluble in acetone,ethanol and methanol.
C1=CC(=C(N=C1)C(=O)O)N
138.126
138.13
98%

Chemical Identifiers

1462-86-8
138.126
BOOMHTFCWOJWFO-UHFFFAOYSA-N
73836
C1=CC(=C(N=C1)C(=O)O)N
C6H6N2O2
MFCD00090153
2-pyridinecarboxylic acid, 3-amino, 2-pyridinecarboxylicacid, 3-amino, 3-amino-2-carboxypyridine, 3-amino-2-picolinic acid, 3-amino-2-pyridinecarboxylic acid, 3-amino-2-pyridinecarboxylicacid, 3-amino-pyridine-2-carboxylic acid, 3-aminopicolinic acid, 3-aminopyridine-2-carboxylic acid, tech., pubchem2015
3-aminopyridine-2-carboxylic acid
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P501c

H315-H317-H319-H335

EINECSNumber : 215-971-8

RTECSNumber : US5175000

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C

Documents