missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopyrazole, 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros AC171030100

 View more versions of this product

Catalog No. AC171030100

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Aminopyrazole
Authentic
100.0
White to light yellow
Oily Liquid or Crystalline Low Melting Solid
≥97.5%
C3H5N3
MFCD00005236
1820-80-0
97.5
Glass bottle
33° to 39°C
10g
98%
83.09
24, 14

Chemical Identifiers

1820-80-0
100.0
C3H5N3
MFCD00005236
≥97.5%
97.5
98%
83.09
3-Aminopyrazole

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.