Please login to your online account to display your discounted pricing

3-Aminopyrazole, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Aminopyrazole
CAS 1820-80-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H5N3
Formula Weight 83.09
MDL Number MFCD00005236
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171030100

  SDS
fisher scientific acros
AC171030100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC171030025

  SDS
fisher scientific acros
171030025
Glass bottle 2.5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Oily Liquid or Crystalline Low Melting Solid
Formula Weight 83.09
Quantity 10g
Color White to light yellow
Melting Point 33° to 39°C
Beilstein 24, 14
CAS Max % 100.0
Chemical Name or Material 3-Aminopyrazole
Molecular Formula C3H5N3
CAS 1820-80-0
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 98%
MDL Number MFCD00005236
CAS Min % 97.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.