missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopyrazole-4-thiocarboxamide, 97%

Chemical Identifiers

CAS 34954-13-7
Molecular Formula C4H6N4S
Molecular Weight (g/mol) 142.18
InChI Key VRWNWVXHINBCSC-UHFFFAOYSA-N
Synonym 3-amino-1h-pyrazole-4-carbothioamide, 3-amino-1h-pyrazole-4-thiocarboxamide, amino 5-aminopyrazol-4-yl methane-1-thione, 1h-pyrazole-4-carbothioamide, 5-amino, 3-amino-2h-pyrazole-4-carbothioamide
PubChem CID 8027117
IUPAC Name 5-amino-1H-pyrazole-4-carbothioamide
SMILES C1=NNC(=C1C(=S)N)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66139MD View Documents Alfa Aesar™
H66139MD
250mg N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH6613903 View Documents Alfa Aesar™
H6613903
1g N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

34954-13-7
142.18
3-amino-1h-pyrazole-4-carbothioamide, 3-amino-1h-pyrazole-4-thiocarboxamide, amino 5-aminopyrazol-4-yl methane-1-thione, 1h-pyrazole-4-carbothioamide, 5-amino, 3-amino-2h-pyrazole-4-carbothioamide
5-amino-1H-pyrazole-4-carbothioamide
C4H6N4S
VRWNWVXHINBCSC-UHFFFAOYSA-N
8027117
C1=NNC(=C1C(=S)N)N

Specifications

Brown to White
34954-13-7
3-amino-1h-pyrazole-4-carbothioamide, 3-amino-1h-pyrazole-4-thiocarboxamide, amino 5-aminopyrazol-4-yl methane-1-thione, 1h-pyrazole-4-carbothioamide, 5-amino, 3-amino-2h-pyrazole-4-carbothioamide
C1=NNC(=C1C(=S)N)N
142.18
142.18
97%
250mg
C4H6N4S
VRWNWVXHINBCSC-UHFFFAOYSA-N
5-amino-1H-pyrazole-4-carbothioamide
8027117
Solid
3-Aminopyrazole-4-thiocarboxamide