Please login to your online account to display your discounted pricing

3-Aminopyrazole-4-carboxylic Acid 95%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  104510010

 View more versions of this product

Catalog No. AC104510010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 127.1
Quantity 1g
Color Almost white to beige
EINECS Number 255-493-7
RTECS Number UQ6390000
TSCA TSCA
Melting Point 121° to 126°C
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00005239
Molecular Formula C4H5N3O2
Packaging Glass bottle
Name Note 95%
Chemical Name or Material 3-Aminopyrazole-4-carboxylic acid
CAS 41680-34-6
Assay Percent Range 95%
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 3-Aminopyrazole-4-carboxylic acid
Name Note 95%
CAS 41680-34-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H5N3O2
Formula Weight 127.1
MDL Number MFCD00005239