Please login to your online account to display your discounted pricing

3-Aminopyrazole-4-carboxylic Acid 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Aminopyrazole-4-carboxylic acid
Name Note 95%
CAS 41680-34-6
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C4H5N3O2
Formula Weight 127.1
MDL Number MFCD00005239
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC104510010

  SDS
thermo fisher scientific
104510010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Crystalline Powder
Formula Weight 127.1
Quantity 1g
Color Almost white to beige
EINECS Number 255-493-7
TSCA TSCA
Melting Point 121° to 126°C
RTECS Number UQ6390000
CAS Min % 94.0
CAS Max % 100.0
MDL Number MFCD00005239
Chemical Name or Material 3-Aminopyrazole-4-carboxylic acid
Name Note 95%
Assay Percent Range 95%
Molecular Formula C4H5N3O2
CAS 41680-34-6
Infrared Spectrum Authentic
Packaging Glass bottle