Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopyrazole-4-carboxamidoxime, 97%

Chemical Identifiers

Chemical Name or Material 3-Aminopyrazole-4-carboxamidoxime
CAS 58046-48-3
Formula Weight 141.13
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6604103

 
alfa aesar™
H6604103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66041MD

 
alfa aesar™
H66041MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 141.13
Chemical Name or Material 3-Aminopyrazole-4-carboxamidoxime
Assay 97%
CAS 58046-48-3
Physical Form Solid
Color White
Quantity 1g