missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropyltriethoxysilane, 99%, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC430948000

 View more versions of this product

Catalog No. AC430948000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

3-Aminopropyltriethoxysilane
Authentic
100.0
1.4200 to 1.4220 (20°C, 589nm)
Clear
800mL
99%
H2N(CH2)3Si(OC2H5)3
MFCD00008207
919-30-2
98.5
AcroSeal™ Glass Bottle
0.9420g/mL
Liquid
≥98.5%
C9H23NO3Si
221.37
3-Triethoxysilylpropylamine

Chemical Identifiers

919-30-2
100.0
C9H23NO3Si
221.37
3-Triethoxysilylpropylamine
≥98.5%
98.5
99%
H2N(CH2)3Si(OC2H5)3
MFCD00008207
3-Aminopropyltriethoxysilane

Safety and Handling

SDS & Safety Documents

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger