missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropyltriethoxysilane, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 151085000

 View more versions of this product

Catalog No. AC151085000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Aminopropyltriethoxysilane
-70.0°C
217.0°C
Authentic
Glass bottle
1.4200 to 1.4220
MFCD00008207,MFCD01324904
04,IV,4273
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
WYTZZXDRDKSJID-UHFFFAOYSA-N
3-triethoxysilylpropan-1-amine
13521
99%
919-30-2
0.9420g/mL
96°C
98.5% min. (GC)
C9H23NO3Si
H2N(CH2)3Si(OC2H5)3
500 g
0.942
Solubility in water: reacts. Other solubilities: soluble in chloroform
CCO[Si](CCCN)(OCC)OCC
221.37
221.37
Liquid

Chemical Identifiers

919-30-2
221.37
WYTZZXDRDKSJID-UHFFFAOYSA-N
13521
CCO[Si](CCCN)(OCC)OCC
C9H23NO3Si
MFCD00008207,MFCD01324904
3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
3-triethoxysilylpropan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 213-048-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only