missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (3-Aminopropyl)diethoxymethylsilane, 97%

Manufacturer:  Alfa Aesar™ L1640122

 View more versions of this product

Catalog No. AAL1640122



Description

Description

Applications
(3-Aminopropyl)diethoxymethylsilane is used as silane coupling agent. The aromatic imine monolayers were prepared by reacting (3-aminopropyl)diethoxymethylsilane.

Solubility
Soluble and reacts with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

(3-Aminopropyl)diethoxymethylsilane
C8H21NO2Si
UN2735
1-propanamine, 3-diethoxymethylsilyl
CCO[Si](C)(CCCN)OCC
191.346
191.35
197°C to 199°C
100g
3179-76-8
MFCD00039785
1744264
HXLAEGYMDGUSBD-UHFFFAOYSA-N
3-[diethoxy(methyl)silyl]propan-1-amine
18511
97%
<-20°C
85°C (185°F)

Chemical Identifiers

3179-76-8
191.346
HXLAEGYMDGUSBD-UHFFFAOYSA-N
18511
CCO[Si](C)(CCCN)OCC
C8H21NO2Si
MFCD00039785
1-propanamine, 3-diethoxymethylsilyl
3-[diethoxy(methyl)silyl]propan-1-amine
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

RTECSNumber : UI0700000

TSCA : TSCA

Documents