Please login to your online account to display your discounted pricing

3-Aminopropionitrile fumarate, 99+%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  104471000

 View more versions of this product

Catalog No. AC104471000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Crystalline Powder
Formula Weight 128.13
Quantity 100g
Color White
TSCA TSCA
EINECS Number 218-208-7
RTECS Number UG0700000
CAS Min % 99.0
CAS Max % 100.0
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
MDL Number MFCD00078321
Chemical Name or Material 3-Aminopropionitrile fumarate
Molecular Formula 0·5 C4H4O4
Linear Formula H2NCH2CH2CN·1/2HO2CCH=CHCO2H
Assay Percent Range 99+%
Name Note 99+%
CAS 2079-89-2
Packaging Plastic bottle

Specifications

Chemical Name or Material 3-Aminopropionitrile fumarate
Name Note 99+%
CAS 2079-89-2
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula 0·5 C4H4O4
Linear Formula H2NCH2CH2CN·1/2HO2CCH=CHCO2H
Formula Weight 128.13
MDL Number MFCD00078321
Accessories

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.

SDS & Safety Documents