missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropionitrile fumarate, 99+%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC104470050

 View more versions of this product

Catalog No. AC104470050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Aminopropionitrile fumarate
Glass bottle
170.0°C to 173.0°C
99+%
99.0
99+%
H2NCH2CH2CN·1/2HO2CCH=CHCO2H
128.13
Crystalline Powder
Authentic
White
5g
2079-89-2
100.0
0·5 C4H4O4
MFCD00078321
128.13
99+%

Chemical Identifiers

2079-89-2
0·5 C4H4O4
128.13
MFCD00078321
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 218-208-7

RTECSNumber : UG0700000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only