missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminopropionitrile, 98%, stab. with potassium carbonate, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1304314

 View more versions of this product

Catalog No. AAA1304314



Description

Description

3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Aminopropionitrile is used for organic synthesis and as pharmaceutical intermediates. As an intermediate used in the manufacture of beta-alanine and pantothenic acid.

Solubility
Soluble in water.

Notes
It is air and moisture sensitive. Incompatible with oxidizing agents, acids and heat.
Specifications

Specifications

3-Aminopropionitrile
151-18-8
79°C to 81°C (16 mmHg)
C3H6N2
H2NCH2CH2CN
25 g
1698848
14,469
Soluble in water.
NCCC#N
70.10
CHEBI:27413
98%
stabilized with potassium carbonate
0.952
Amine-like
1.4375
MFCD00014820
UN2735
Air and moisture sensitive
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
AGSPXMVUFBBBMO-UHFFFAOYSA-N
3-aminopropanenitrile
1647
70.1

Chemical Identifiers

151-18-8
70.10
AGSPXMVUFBBBMO-UHFFFAOYSA-N
1647
3-aminopropanenitrile
C3H6N2
MFCD00014820
3-aminopropionitrile, 2-cyanoethylamine, aminopropionitrile, beta-aminopropionitrile, bapn, 3-aminopropiononitrile, beta-cyanoethylamine, propanenitrile, 3-amino, beta-alaninenitrile, propionitrile, 3-amino
CHEBI:27413
NCCC#N
Safety and Handling

Safety and Handling

GHS H Statement
H301-H314-H318-H361-H227
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
Combustible liquid.

P201-P202-P210-P235-P260-P264b-P270-P271-P281-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H227-H302-H314-H335-H361

DOTInformation : Hazard Class: 8; Packaging Group: II

EINECSNumber : 205-786-0

RTECSNumber : UG0350000

TSCA : Yes

Recommended Storage : Keep cold; Store under Argon

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only