missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Aminopropane-1-sulfonic acid, 97%

Manufacturer:  Alfa Aesar™ H5591803

 View more versions of this product

Catalog No. AAH5591803



Specifications

Specifications

3-Aminopropane-1-sulfonic acid
1g
C3H9NO3S
MFCD00008225
1-propanesulfonic acid, 3-amino
C(CN)CS(=O)(=O)O
139.169
CHEBI:1457
97%
293°C (decomposition)
3687-18-1
H2N(CH2)3SO3H
14,10218
SNKZJIOFVMKAOJ-UHFFFAOYSA-N
3-aminopropane-1-sulfonic acid
1646
139.17

Chemical Identifiers

3687-18-1
139.169
SNKZJIOFVMKAOJ-UHFFFAOYSA-N
1646
3-aminopropane-1-sulfonic acid
C3H9NO3S
MFCD00008225
1-propanesulfonic acid, 3-amino
CHEBI:1457
C(CN)CS(=O)(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-977-4

TSCA : No

Recommended Storage : Ambient temperatures

Documents