missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Aminophthalhydrazide, 98%, pure, ACROS Organics™

3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0

Manufacturer:  Thermo Fisher Scientific AC153851000

 View more versions of this product

Catalog No. AC153851000



Description

Description

3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0

Specifications

Specifications

3-Aminophthalhydrazide
Glass bottle
319.0°C to 320.0°C
98%
97.5
98%
MFCD00006890
15,566
(2% in 1 N NaOH) Clear yellow to yellow-green
C1=CC2=C(C(=C1)N)C(=O)NNC2=O
177.16
177.16
98%
Authentic
Blue-Purple to Green
100g
521-31-3
100.0
C8H7N3O2
25,IV,4192
1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalhydrazide, 3-aminophthalic acid hydrazide, 3-aminophthalic hydrazide, 3-aminophthalylhydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, 5-amino-2,3-dihydro-1,4-phthalazinedione, ccris 5962, luminol, unii-5exp385q4f
HWYHZTIRURJOHG-UHFFFAOYSA-N
5-amino-2,3-dihydrophthalazine-1,4-dione
10638
Powder
Pure

Chemical Identifiers

521-31-3
177.16
HWYHZTIRURJOHG-UHFFFAOYSA-N
10638
C1=CC2=C(C(=C1)N)C(=O)NNC2=O
C8H7N3O2
MFCD00006890
1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalhydrazide, 3-aminophthalic acid hydrazide, 3-aminophthalic hydrazide, 3-aminophthalylhydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, 5-amino-2,3-dihydro-1,4-phthalazinedione, ccris 5962, luminol, unii-5exp385q4f
5-amino-2,3-dihydrophthalazine-1,4-dione
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue

GHS Signal Word: Warning

EINECSNumber : 208-309-4

RTECSNumber : TH8890060

TSCA : TSCA

SDS Documents