Learn More
3-Aminophthalhydrazide, 98%, pure, Thermo Scientific Chemicals
3-Aminophthalhydrazide, 98%, pure, Acros Organics, Quantity: 25g, Packaging: Glass bottle, Blue-Purple to Green, Grade: Pure, Melting Point: 319.0 deg.C to 320.0 deg.C, Molecular Weight: 177.16, Percent Purity: 98%, Assay Percent Range: 98%, Beilstein: 25,IV,4192, CAS: 521-31-3, CAS Max %: 100.0
Supplier: Thermo Scientific Chemicals 153850050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
3-Aminophthalhydrazide | |
521-31-3 | |
100.0 | |
Blue-Purple to Green | |
98% | |
C8H7N3O2 | |
5 g | |
15,566 | |
(2% in 1 N NaOH) Clear yellow to yellow-green | |
C1=CC2=C(C(=C1)N)C(=O)NNC2=O | |
177.16 | |
177.16 | |
Pure |
98% | |
97.5 | |
319.0°C to 320.0°C | |
Authentic | |
Glass bottle | |
MFCD00006890 | |
25,IV,4192 | |
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide | |
HWYHZTIRURJOHG-UHFFFAOYSA-N | |
5-amino-2,3-dihydrophthalazine-1,4-dione | |
10638 | |
98% | |
Powder |
Chemical Identifiers
521-31-3 | |
177.16 | |
HWYHZTIRURJOHG-UHFFFAOYSA-N | |
10638 | |
C1=CC2=C(C(=C1)N)C(=O)NNC2=O |
C8H7N3O2 | |
MFCD00006890 | |
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide | |
5-amino-2,3-dihydrophthalazine-1,4-dione |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue
GHS Signal Word: Warning
EINECSNumber : 208-309-4
RTECSNumber : TH8890060
TSCA : TSCA
RUO – Research Use Only