Learn More
3-Aminophthalhydrazide, 98%, Thermo Scientific Chemicals
Alfa Aesar 3-Aminophthalhydrazide, 98%, Quantity: 25g, Melting Point: 325 deg.C to 330 deg.C, Molecular Weight: 177.163, Percent Purity: 98%, Beilstein: 383929, CAS: 521-31-3, EINECS Number: 208-309-4, Formula Weight: 177.16, Health Hazard 1: H315-H319-H335
Supplier: Thermo Scientific Chemicals A1459714
Description
3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Aminophthalhydrazide is used in the detection of copper, iron, peroxides and cyanides. It exhibits chemiluminescence and utilized to measure opsonic and phagocytic function. It acts as a biological sensor and involved in the detection of polymorphonuclear leukocytes response in a patient with a myeloperoxidase deficiency. Further, it is used as a forensic test for blood.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and strong reducing agents.
Specifications
3-Aminophthalhydrazide | |
325°C to 330°C | |
C8H7N3O2 | |
25 g | |
14,5600 | |
Soluble in water. | |
C1=CC2=C(C(=C1)N)C(=O)NNC2=O | |
177.163 | |
177.16 |
521-31-3 | |
Odorless | |
MFCD00006890 | |
383929 | |
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide | |
HWYHZTIRURJOHG-UHFFFAOYSA-N | |
5-amino-2,3-dihydrophthalazine-1,4-dione | |
10638 | |
98% |
Chemical Identifiers
521-31-3 | |
177.163 | |
HWYHZTIRURJOHG-UHFFFAOYSA-N | |
10638 | |
C1=CC2=C(C(=C1)N)C(=O)NNC2=O |
C8H7N3O2 | |
MFCD00006890 | |
luminol, 3-aminophthalhydrazide, 3-aminophthalic hydrazide, 5-amino-2,3-dihydro-1,4-phthalazinedione, 1,4-phthalazinedione, 5-amino-2,3-dihydro, 3-aminophthalic acid hydrazide, 5-amino-1,2,3,4-tetrahydrophthalazine-1,4-dione, unii-5exp385q4f, ccris 5962, 3-aminophthalylhydrazide | |
5-amino-2,3-dihydrophthalazine-1,4-dione |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-309-4
RTECSNumber : TH8890060
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only