Please login to your online account to display your discounted pricing

3-Aminophenylboronic acid hydrochloride, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Aminophenylboronic acid hydrochloride
CAS 85006-23-1
CAS Min % 97.5
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98%
Molecular Formula C6H8BNO2·ClH
Formula Weight 173.41
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC435530010

  SDS
thermo fisher scientific
AC435530010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Crystalline Powder
Formula Weight 173.41
Quantity 1g
Color Off-white to gray
CAS Max % 100.0
Molecular Formula C6H8BNO2·ClH
CAS 85006-23-1
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS Min % 97.5
Assay Percent Range 98%
Chemical Name or Material 3-Aminophenylboronic acid hydrochloride
Infrared Spectrum Authentic
Elemental Analysis 40.52 to 42.60% C, 7.88 to 8.28% N
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.